Learn More
Medchemexpress LLC Duocarmycin GA | 1613286-59-1 | 99.0% | 476.95 g/mol | C26H25ClN4O3 | 1 MG
SDP
Catalog No. 5000203461
Encompass_Preferred
Duocarmycin GA is a DNA-alkylating cytotoxin used as a research-grade antibody-drug conjugate (ADC) payload that binds in the minor groove of DNA and is active against multi-drug-resistant cell lines.
  • High purity of 99.0%.
  • Molecular weight 476.95 g/mol and formula C26H25ClN4O3.
  • Available in mg-scale quantities suitable for conjugation and in vitro studies.
  • Solid form, light yellow to yellow, facilitating handling for synthesis workflows.
  • Store protected from light under nitrogen at -20°C; in solvent, store at -80°C up to 6 months.
Catalog No. 50-002-03461 Supplier Medchemexpress LLC Supplier No. HY1288731MG
Duocarmycin GA is a DNA-alkylating cytotoxin used as a research-grade antibody-drug conjugate (ADC) payload that binds in the minor groove of DNA and is active against multi-drug-resistant cell lines.
  • High purity of 99.0%.
  • Molecular weight 476.95 g/mol and formula C26H25ClN4O3.
  • Available in mg-scale quantities suitable for conjugation and in vitro studies.
  • Solid form, light yellow to yellow, facilitating handling for synthesis workflows.
  • Store protected from light under nitrogen at -20°C; in solvent, store at -80°C up to 6 months.