Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Duocarmycin GA | 1613286-59-1 | 99.0% | 476.95 g/mol | C26H25ClN4O3 | 5 MG
SDP

Catalog No. 5000203654
Encompass_Preferred

Duocarmycin GA is a DNA minor-groove alkylating agent used as an antibody-drug conjugate (ADC) payload and research cytotoxin; it is active against some multi-drug resistant cell lines.

  • Purity 99.0% suitable for research applications.
  • Molecular formula C26H25ClN4O3 and molecular weight 476.95 g/mol.
  • CAS number 1613286-59-1 for unambiguous identification.
  • Available in small packaging sizes for ADC development and in vitro research, including 5 MG.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-03654 Supplier Medchemexpress LLC Supplier No. HY1288735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Duocarmycin GA is a DNA minor-groove alkylating agent used as an antibody-drug conjugate (ADC) payload and research cytotoxin; it is active against some multi-drug resistant cell lines.

  • Purity 99.0% suitable for research applications.
  • Molecular formula C26H25ClN4O3 and molecular weight 476.95 g/mol.
  • CAS number 1613286-59-1 for unambiguous identification.
  • Available in small packaging sizes for ADC development and in vitro research, including 5 MG.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.