Learn More
Sigma Aldrich Fine Chemicals Biosciences DuolinkR In Situ Detection
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DUO9201430RXN
Encompass
Duolink(R) In Situ Detection Reagents Green contains all the necessary Duolink In situ reagents to perform the amplification and detection of bound PLA(R) probes. The detection probes contain a fluorophore (lex 495 nm and lem 527 nm) which may be visualized using the same filter as Cy(R)2 or fITC. Experiments conducted using Duolink In situ reagents can detect and visualize protein interactions protein expression levels and post translational modifications at the single molecule level in fixed cells and tissue samples.
Catalog No. 50-290-6057