Learn More
Sigma Aldrich Fine Chemicals Biosciences DuolinkR In Situ PLAR Prob
Catalog No. 502905953
Encompass
Duolink(R) In Situ PLA(R) Probe Anti-Mouse MINUS contains affinity purified donkey anti-mouse IgG (HL) which reacts with whole molecule mouse IgG and also reacts with the light chains of other mouse immunoglobulins.
Catalog No. 50-290-5953 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DUO9200430RXN
Duolink(R) In Situ PLA(R) Probe Anti-Mouse MINUS contains affinity purified donkey anti-mouse IgG (HL) which reacts with whole molecule mouse IgG and also reacts with the light chains of other mouse immunoglobulins.