Learn More
Fine Science Tools Dumont SS Fine Forceps, Straight, 0.1 x 0.06mm, 13.5 cm
SDP
Catalog No. NC9310636
Encompass
Style: SS/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumoxel/ Length: 13.5cm
Catalog No. NC9310636 Supplier Fine Science Tools Supplier No. 1120033
Style: SS/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumoxel/ Length: 13.5cm