Learn More
Biomatik Corporation Duox2 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10887
Encompass_Preferred
Duox2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to DUOX2. The Duox2 Polyclonal Antibody has been validated for the following applications: ELISA,WB,IHC,IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dual Oxidase 2 Protein (1-100Aa). Applications: ELISA,WB,IHC,IF. Target Synonyms: DUOX2. Accession Number: Q9NRD8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7688