Learn More
TARGETMOL CHEMICALS INC DUP-697 5MG
SDP
Catalog No. 502581246
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DuP-697 is an irreversible and specific inhibitor of COX-2 with IC50 values of 10 nM and 800 nM for human COX-2 and COX-1. DuP-697 shows antiproliferative activity with an IC50 of 42.8 nM. DuP-697 exhibits antiangiogenic anti-inflammatory antipyretic and apoptotic effects. purity: 100%
Catalog No. 50-258-1246 Supplier TARGETMOL CHEMICALS INC Supplier No. T151815MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DuP-697 is an irreversible and specific inhibitor of COX-2 with IC50 values of 10 nM and 800 nM for human COX-2 and COX-1. DuP-697 shows antiproliferative activity with an IC50 of 42.8 nM. DuP-697 exhibits antiangiogenic anti-inflammatory antipyretic and apoptotic effects. purity: 100%