Learn More
Apexbio Technology LLC DuP 697 88149-94-4 50mg
SDP
Catalog No. 5000091668
Encompass_Preferred
DuP 697 (CAS 88149-94-4) is an ortho-diaryl heterocyclic compound that selectively and irreversibly inhibits cyclooxygenase-2 (COX-2) displaying an IC50 of 10 nM for human COX-2 and 800 nM for COX-1 DuP 697 is demonstrated to suppress prostaglandin synthesis and in vitro exhibits antiproliferative pro-apoptotic and anti-angiogenic effects in HT29 colorectal cancer cells (IC50 42 8 nM) This small molecule is commonly utilized to explore COX-2-mediated inflammatory pathways as well as tumor biology providing a valuable tool for dissecting mechanisms underlying inflammation and cancer
Catalog No. 50-000-91668 Supplier Apexbio Technology LLC Supplier No. B673750
DuP 697 (CAS 88149-94-4) is an ortho-diaryl heterocyclic compound that selectively and irreversibly inhibits cyclooxygenase-2 (COX-2) displaying an IC50 of 10 nM for human COX-2 and 800 nM for COX-1 DuP 697 is demonstrated to suppress prostaglandin synthesis and in vitro exhibits antiproliferative pro-apoptotic and anti-angiogenic effects in HT29 colorectal cancer cells (IC50 42 8 nM) This small molecule is commonly utilized to explore COX-2-mediated inflammatory pathways as well as tumor biology providing a valuable tool for dissecting mechanisms underlying inflammation and cancer