Learn More
Medchemexpress LLC HY-103591 100mg Medchemexpress, DUPA(OtBu)-OH CAS:1026987-94-9 Purity:>98%
SDP
Catalog No. 501968132
Encompass_Preferred
Medchemexpress, HY-103591 100mg DUPA(OtBu)-OH CAS:1026987-94-9 DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8132 Supplier Medchemexpress LLC Supplier No. HY103591100MG
Medchemexpress, HY-103591 100mg DUPA(OtBu)-OH CAS:1026987-94-9 DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.