Learn More
Medchemexpress LLC Dupa(OtBu)-OH | 1026987-94-9 | 98.0% | 488.57 g/mol | C23H40N2O9 | 25 MG
SDP
Catalog No. 5000244610
Encompass_Preferred
DUPA(OtBu)-OH is a protected DUPA precursor used in the synthesis of prostate-specific membrane antigen (PSMA) targeting ligands and drug conjugates for research applications.
  • Precursor for DUPA-based PSMA-targeting ligands and conjugates
  • Includes tert-butyl (OtBu) protection for carboxyl functionality
  • Useful for synthesizing docetaxel and paclitaxel conjugates
  • High purity suitable for research applications (98.0%)
  • Available in multiple sizes including 25 mg
Catalog No. 50-002-44610 Supplier Medchemexpress LLC Supplier No. HY10359125MG
DUPA(OtBu)-OH is a protected DUPA precursor used in the synthesis of prostate-specific membrane antigen (PSMA) targeting ligands and drug conjugates for research applications.
  • Precursor for DUPA-based PSMA-targeting ligands and conjugates
  • Includes tert-butyl (OtBu) protection for carboxyl functionality
  • Useful for synthesizing docetaxel and paclitaxel conjugates
  • High purity suitable for research applications (98.0%)
  • Available in multiple sizes including 25 mg