Learn More
Cayman Chemical DUPA(OtBu)-OH, Purity: ≥98%, CAS: 1026987-94-9, 50 mg
SDP
Catalog No. 502917140
Encompass_Preferred
A DUPA-conjugate precursor; has been used in the synthesis of PSMA-targeted radio- and optical imaging agents and prodrugs with anticancer activity
Catalog No. 50-291-7140 Supplier Cayman Chemical Supplier No. 3961750 MG
A DUPA-conjugate precursor; has been used in the synthesis of PSMA-targeted radio- and optical imaging agents and prodrugs with anticancer activity