Learn More
Medchemexpress LLC HY-P9926 5mg , Dupilumab CAS:1190264-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99265MG
Encompass_Preferred
HY-P9926 5mg Dupilumab CAS: 1190264-60-8 Dupilumab (REGN-668) is a fully human mAb to IL-4 receptor α (IL-4Rα) that inhibits both IL-4 and IL-13 signaling, markedly improved moderate-to-severe atopic dermatitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1222