Learn More
Grainger Little Giant Duplex Alternator, NEMA 4, For Inline Systems
Catalog No. 502773126
Encompass_Preferred
This duplex alternator is designed to facilitate the operation of multiple pumps in parallel, supporting applications with larger volume demands. It is compatible with the entire SubDrive product family and provides essential control and monitoring features for inline constant pressure systems.
  • Selectable alternate timing allows for efficient pump load distribution
  • Status indicator lights and alarm circuit contacts provide real-time monitoring
  • Integrated fault detection and fail-safe operation enhance system reliability
  • Manual override push-button enables direct control when needed
  • NEMA 4 enclosure ensures protection for various operating environments
  • Includes controller unit, pressure sensor cables, SubDrive cables, mounting brackets, transformer, and installation manual
Catalog No. 50-277-3126 Supplier Grainger Supplier No. 13T431
This duplex alternator is designed to facilitate the operation of multiple pumps in parallel, supporting applications with larger volume demands. It is compatible with the entire SubDrive product family and provides essential control and monitoring features for inline constant pressure systems.
  • Selectable alternate timing allows for efficient pump load distribution
  • Status indicator lights and alarm circuit contacts provide real-time monitoring
  • Integrated fault detection and fail-safe operation enhance system reliability
  • Manual override push-button enables direct control when needed
  • NEMA 4 enclosure ensures protection for various operating environments
  • Includes controller unit, pressure sensor cables, SubDrive cables, mounting brackets, transformer, and installation manual