Learn More
Bioworld Duplex Buffer, 4 L
Catalog No. 50196826
Encompass_Preferred
Product Description Duplex buffer contains 100 mM potassium acetate, 30 mM HEPES and has a pH level of 7.5. It comes as a sterile solution in DEPC-treated water and is for initial resuspension, annealing and storage of duplex DsiRNA products. The salt is necessary for oligonucleotide hybridization.
Catalog No. 50-196-826 Supplier Bioworld Supplier No. 107500084
Product Description Duplex buffer contains 100 mM potassium acetate, 30 mM HEPES and has a pH level of 7.5. It comes as a sterile solution in DEPC-treated water and is for initial resuspension, annealing and storage of duplex DsiRNA products. The salt is necessary for oligonucleotide hybridization.