Learn More
New England Biolabs, Inc. Duplex DNase - 1000 units
SDP
Catalog No. 502551931
Encompass
An engineered double-strand specific DNA endonuclease, dsDNase preferentially degrades double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA) or RNA. This specificity toward dsDNA allows the enzyme to degrade contaminating dsDNA while maintaining the integrity of single stranded DNA molecules such as primers and probes within PCR mixes, allowing for efficient decontamination of PCR master mixes. Moreover, dsDNase displays the unique property of cleaving the DNA strand of a DNA:RNA hybrid duplex.
Catalog No. 50-255-1931 Supplier New England Biolabs, Inc. Supplier No. M7635S
An engineered double-strand specific DNA endonuclease, dsDNase preferentially degrades double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA) or RNA. This specificity toward dsDNA allows the enzyme to degrade contaminating dsDNA while maintaining the integrity of single stranded DNA molecules such as primers and probes within PCR mixes, allowing for efficient decontamination of PCR master mixes. Moreover, dsDNase displays the unique property of cleaving the DNA strand of a DNA:RNA hybrid duplex.