Learn More
MSC DuPont Bib Overall and Jacket, Chemical Resistant, Durable, Tan, 6/Case
Catalog No. 5000781594
Encompass
This bib overall and jacket combo is designed for chemical resistance and durability in hazardous environments, offered in tan for discretionary use.
  • Features multi-layer film barrier fabric laminated to a durable 2.0 oz/yd² polypropylene substrate.
  • Includes mandarin collar, jam fit cuffs, storm flap, open ankles, and adjustable bib overall straps.
  • Taped seams offer chemical resistance equal to or greater than the base fabric.
  • Jacket has a front zipper with a double storm flap and hook and loop closure.
  • Comes with glove inserts for user-supplied gloves.
  • Manufactured without natural rubber latex; meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).
  • Provides heavy-duty protection against a wide variety of chemical hazards and is strong for rigorous activities and liquid splash.
Catalog No. 50-007-81594 Supplier MSC Supplier No. 55525547
This bib overall and jacket combo is designed for chemical resistance and durability in hazardous environments, offered in tan for discretionary use.
  • Features multi-layer film barrier fabric laminated to a durable 2.0 oz/yd² polypropylene substrate.
  • Includes mandarin collar, jam fit cuffs, storm flap, open ankles, and adjustable bib overall straps.
  • Taped seams offer chemical resistance equal to or greater than the base fabric.
  • Jacket has a front zipper with a double storm flap and hook and loop closure.
  • Comes with glove inserts for user-supplied gloves.
  • Manufactured without natural rubber latex; meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).
  • Provides heavy-duty protection against a wide variety of chemical hazards and is strong for rigorous activities and liquid splash.