Learn More
Cardinal Health™ Blue Premium Bouffant Cap
Catalog No. 19160275 Shop All Cardinal Health Products
Change view
Click to view available options
24 in. Blue
Catalog No.
19-160-275 24 in. Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-160-275 Supplier Cardinal Health™ Supplier No. B24HD
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to provide strength and fluid-resistance while offering breathability and comfort.
  • Constructed of heavyweight spunbonded polypropylene
  • 100% latex-free
  • Synthetic elastic conforms to head for a personalized fit
  • 24 in. diameter
Order Info
Packaged 75 per box, 10 boxes per case.
Specifications
Disposable Single-use
Diameter (Metric) 61 cm