Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Cool-Guard™ Personal Cooling Vest

Catalog No. 19088787 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Cooling Vest; X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-787 Cooling Vest; X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-088-787 Supplier DuPont™ Supplier No. 996000BU00000100

missing translation for 'validMainframeMsg'

State-of-the-art cooling technology maintains proper body temperature

The Cool-Guard vest provides cooling without the discomfort, vasoconstriction, or moisture buildup of typical icepack systems.

  • Phase-change gel packs absorb heat generated by the body
  • Provide 1.5 to 2 hours of cooling at 43.3°C (110°F)
  • Gel packs recharge quickly in ice water within 20 minutes
  • Ergonomically designed blue Banox™ vest is light and flexible
  • Fitted with adjustable straps so one size fits all
  • Flame-retardant

Order Info

Total weight (including phase-change packs): 3.88 lb. (1.8kg)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Vest
Size X-Large
Color Blue
For Use With (Application) Aids in reducing potential for heat stress while wearing chemical protective garments
Includes One Set of 4 Phase Change Packs
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.