Learn More
MSC DuPont Coverall, Disposable, Flame Resistant, Hood, Elastic Cuffs, 25/Case
Catalog No. 5000795217
Encompass
DuPont ProShield 6 SFR Coveralls are lightweight, disposable garments designed for use as an overgarment in environments with potential flame or electric arc exposure. They help protect primary flame-resistant apparel from contamination.
  • Standard fit hood
  • Stormflap
  • Elastic wrists and ankles
  • Serged seams
  • Pin lock slider zipper pull
  • Front zipper closure
  • Resists chemical, flame, and tear
  • Made from PROSHIELD® 6SFR fabric
  • Fabric weight 2.4 oz/yd²
Catalog No. 50-007-95217 Supplier MSC Supplier No. 75689372
DuPont ProShield 6 SFR Coveralls are lightweight, disposable garments designed for use as an overgarment in environments with potential flame or electric arc exposure. They help protect primary flame-resistant apparel from contamination.
  • Standard fit hood
  • Stormflap
  • Elastic wrists and ankles
  • Serged seams
  • Pin lock slider zipper pull
  • Front zipper closure
  • Resists chemical, flame, and tear
  • Made from PROSHIELD® 6SFR fabric
  • Fabric weight 2.4 oz/yd²