Learn More
MSC Dupont Coverall, Tychem 6000, Large, Gray, Hooded, Taped Seams, 6/Case
Catalog No. 5000796231
Encompass
The Tychem 6000 hooded coverall with attached socks is strong and durable, offering a broad chemical barrier against chemical warfare agents and industrial chemicals. This gray coverall features a respirator fit hood, elastic wrists, attached socks, a double storm flap, a semi-auto locking slider zipper pull, and taped seams.
  • Constructed from a proprietary barrier film laminated to a Tyvek substrate
  • Provides at least 30 minutes of protection against over 180 challenge chemicals
  • Strong, durable, and lightweight design
  • Features a three-piece hood for respirator compatibility
  • Equipped with a storm flap and adhesive closure over the zipper
  • Includes integrated socks and elastic wrists for a secure fit
  • Manufactured without natural rubber latex (as of January 2023)
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
Catalog No. 50-007-96231 Supplier MSC Supplier No. 02144731
The Tychem 6000 hooded coverall with attached socks is strong and durable, offering a broad chemical barrier against chemical warfare agents and industrial chemicals. This gray coverall features a respirator fit hood, elastic wrists, attached socks, a double storm flap, a semi-auto locking slider zipper pull, and taped seams.
  • Constructed from a proprietary barrier film laminated to a Tyvek substrate
  • Provides at least 30 minutes of protection against over 180 challenge chemicals
  • Strong, durable, and lightweight design
  • Features a three-piece hood for respirator compatibility
  • Equipped with a storm flap and adhesive closure over the zipper
  • Includes integrated socks and elastic wrists for a secure fit
  • Manufactured without natural rubber latex (as of January 2023)
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)