Learn More
MSC Dupont Coveralls, 4X-Large, Tychem 5000, Taped Seam, 6/Pack
Catalog No. 5000782799
Encompass
These chemical-resistant coveralls are designed for light-duty, hazardous material applications. They feature taped seams, a zipper closure, and elastic cuffs, providing protection against various hazards.
  • Collared design with elastic cuffs and attached socks
  • Taped seams for chemical resistance
  • Constructed from Tychem 5000 material
  • Zipper closure with hook-and-loop storm flap
  • Attached dual-layer gloves (multi-layer laminate internal, neoprene external) and outer boot flaps
  • Liquid, dust, and reduced spray tight protection
  • Meets EPA level B and 16 CFR 1610 Class 1 flame resistance
  • Protection against bloodborne pathogens, hazardous particulates, and chemical splashes/sprays
  • Certified to ASTM F1670 and F1671 standards
  • Non-sterile, non-cleanroom, light-duty application
Catalog No. 50-007-82799 Supplier MSC Supplier No. 68636216
These chemical-resistant coveralls are designed for light-duty, hazardous material applications. They feature taped seams, a zipper closure, and elastic cuffs, providing protection against various hazards.
  • Collared design with elastic cuffs and attached socks
  • Taped seams for chemical resistance
  • Constructed from Tychem 5000 material
  • Zipper closure with hook-and-loop storm flap
  • Attached dual-layer gloves (multi-layer laminate internal, neoprene external) and outer boot flaps
  • Liquid, dust, and reduced spray tight protection
  • Meets EPA level B and 16 CFR 1610 Class 1 flame resistance
  • Protection against bloodborne pathogens, hazardous particulates, and chemical splashes/sprays
  • Certified to ASTM F1670 and F1671 standards
  • Non-sterile, non-cleanroom, light-duty application