Learn More
Enterprise Technology Solutions Dupont Coveralls, Non-Woven, Zipper, Large, 25/CT, White
SDP
Catalog No. 5000828141
Encompass_Preferred
Tyvek coveralls provide a balance of protection, comfort and durability with a wide range of movement while stretching and bending. Set-sleeve pattern allows greater arm movement. Unique, nonwoven technology of the coveralls delivers a comfortable, inherent barrier protection against dry particulate hazards and light liquid splash. Lightweight, low-linting, antistatic flashspun polyolefin stops sub-micron particles better than reusable garments, even after abrasion. Coveralls also feature a zipper front with a nonsparking zipper, collar, no elastic and serged seams that are strong and stress-resistant.
Catalog No. 50-008-28141 Supplier Enterprise Technology Solutions Supplier No. DUP120SWHLG00
Tyvek coveralls provide a balance of protection, comfort and durability with a wide range of movement while stretching and bending. Set-sleeve pattern allows greater arm movement. Unique, nonwoven technology of the coveralls delivers a comfortable, inherent barrier protection against dry particulate hazards and light liquid splash. Lightweight, low-linting, antistatic flashspun polyolefin stops sub-micron particles better than reusable garments, even after abrasion. Coveralls also feature a zipper front with a nonsparking zipper, collar, no elastic and serged seams that are strong and stress-resistant.