Learn More
MSC Dupont Disposable Coverall, ProShield 50, Liquid/Splash, White, Medium, 25/Case
Catalog No. 5000796188
Encompass
The ProShield 50 Coverall is a lightweight, microporous film garment designed to provide a barrier against non-hazardous aerosols, light liquid splash, and dry particles. It is suitable for janitorial, sanitation, and general industrial maintenance applications.
  • Constructed from ProShield 50, a composite of microporous film and nonwoven fabric
  • Coverall design with collar, front zipper closure, open wrists, and open ankles
  • Features serged seams for enhanced durability
  • Lightweight and roomy design offers greater comfort and mobility
  • Manufactured without natural rubber latex components
Catalog No. 50-007-96188 Supplier MSC Supplier No. 06589618
The ProShield 50 Coverall is a lightweight, microporous film garment designed to provide a barrier against non-hazardous aerosols, light liquid splash, and dry particles. It is suitable for janitorial, sanitation, and general industrial maintenance applications.
  • Constructed from ProShield 50, a composite of microporous film and nonwoven fabric
  • Coverall design with collar, front zipper closure, open wrists, and open ankles
  • Features serged seams for enhanced durability
  • Lightweight and roomy design offers greater comfort and mobility
  • Manufactured without natural rubber latex components