Learn More
MSC Dupont Disposable Coveralls, Chemical Protection, 2X-Large, Tychem 5000, 6 Pack
Catalog No. 5000802778
Encompass
These disposable coveralls offer comprehensive protection against chemical, liquid, and splash hazards. Constructed from Tychem 5000 material, they are designed for durability in rigorous industrial applications.
  • Protects against liquid, chemical, and splash hazards
  • Features zipper and stormflaps closure
  • Taped seams for enhanced protection
  • Elastic cuffs and ankles for secure fit
  • Made from Tychem 5000 material
  • Suitable for industrial manufacturing, chemical handling, and hazardous waste clean-up
Catalog No. 50-008-02778 Supplier MSC Supplier No. 66598095
These disposable coveralls offer comprehensive protection against chemical, liquid, and splash hazards. Constructed from Tychem 5000 material, they are designed for durability in rigorous industrial applications.
  • Protects against liquid, chemical, and splash hazards
  • Features zipper and stormflaps closure
  • Taped seams for enhanced protection
  • Elastic cuffs and ankles for secure fit
  • Made from Tychem 5000 material
  • Suitable for industrial manufacturing, chemical handling, and hazardous waste clean-up