Learn More
MSC Dupont Disposable Coveralls, Liquid Splash Protection, Medium, ProShield 50, 25/Pk.
Catalog No. 5000673621
Encompass
ProShield® 50 Microporous Film Garments provide a barrier against non-hazardous aerosols, light liquid splash, and dry particles. These garments are lightweight, and their roomy design allows for greater comfort and mobility.
  • Provides barrier against non-hazardous aerosols, light liquid splash, and dry particles
  • Lightweight and roomy design for comfort and mobility
  • Features zipper closure, serged seams, and open cuffs and ankles
  • Suitable for janitorial, sanitation, industrial maintenance, asbestos abatement, and hazardous remediation
  • Material weight 1.5000 oz.
  • Meets standards including ASTM D751, ASTM D5034, AATCC 127
  • ISO Class non-cleanroom
  • Color: White
Catalog No. 50-006-73621 Supplier MSC Supplier No. 54753678
ProShield® 50 Microporous Film Garments provide a barrier against non-hazardous aerosols, light liquid splash, and dry particles. These garments are lightweight, and their roomy design allows for greater comfort and mobility.
  • Provides barrier against non-hazardous aerosols, light liquid splash, and dry particles
  • Lightweight and roomy design for comfort and mobility
  • Features zipper closure, serged seams, and open cuffs and ankles
  • Suitable for janitorial, sanitation, industrial maintenance, asbestos abatement, and hazardous remediation
  • Material weight 1.5000 oz.
  • Meets standards including ASTM D751, ASTM D5034, AATCC 127
  • ISO Class non-cleanroom
  • Color: White