Learn More
MSC Dupont Disposable Coveralls, Particle Protection, 2X-Large, Tyvek 400, 25/Pk.
Catalog No. 5000687012
Encompass
This garment is made from Tyvek® 400, a unique, nonwoven material composed of flash-spun high-density polyethylene. It offers an optimal balance of protection, durability, and comfort for a limited-use fabric technology.
  • Provides an inherent barrier against particles as small as 1.0 micron.
  • Protection is integrated directly into the fabric, with no films or laminates to abrade or wear away.
  • Offers durability advantage over microporous film fabrics, ensuring better barrier performance even after wear and abrasion.
  • Suitable for applications including lead and asbestos abatement/remediation, general clean-up, general maintenance/operations, and spray painting.
Catalog No. 50-006-87012 Supplier MSC Supplier No. 41551276
This garment is made from Tyvek® 400, a unique, nonwoven material composed of flash-spun high-density polyethylene. It offers an optimal balance of protection, durability, and comfort for a limited-use fabric technology.
  • Provides an inherent barrier against particles as small as 1.0 micron.
  • Protection is integrated directly into the fabric, with no films or laminates to abrade or wear away.
  • Offers durability advantage over microporous film fabrics, ensuring better barrier performance even after wear and abrasion.
  • Suitable for applications including lead and asbestos abatement/remediation, general clean-up, general maintenance/operations, and spray painting.