Learn More
MSC Dupont Disposable Coveralls, Tychem 4000, 5X-Large, White, 12/Pk.
Catalog No. 5000677862
Encompass
These disposable coveralls offer effective protection against a range of chemical environments and bloodborne pathogens. They are constructed from Tychem 4000 material, designed for rugged durability and abrasion resistance.
  • Delivers effective protection against a range of chemical environments
  • Utilizes Saranex 23-P film laminated to Tyvek fabric
  • Provides rugged and durable fabric that resists abrasion
  • Offers at least 30 minutes of protection against more than 124 chemical challenges
  • Features zipper closure with stormflaps
  • Includes elastic cuffs and attached socks
  • Bound seams for enhanced protection
  • Suitable for chemical mixing, hazardous remediation, and emergency medical response
  • Meets OSHA 16 CFR 1610, ASTM D5034, ASTM D3786, ASTM D3776, ASTM D1777 standards
Catalog No. 50-006-77862 Supplier MSC Supplier No. 06838692
These disposable coveralls offer effective protection against a range of chemical environments and bloodborne pathogens. They are constructed from Tychem 4000 material, designed for rugged durability and abrasion resistance.
  • Delivers effective protection against a range of chemical environments
  • Utilizes Saranex 23-P film laminated to Tyvek fabric
  • Provides rugged and durable fabric that resists abrasion
  • Offers at least 30 minutes of protection against more than 124 chemical challenges
  • Features zipper closure with stormflaps
  • Includes elastic cuffs and attached socks
  • Bound seams for enhanced protection
  • Suitable for chemical mixing, hazardous remediation, and emergency medical response
  • Meets OSHA 16 CFR 1610, ASTM D5034, ASTM D3786, ASTM D3776, ASTM D1777 standards