Learn More
Cardinal Health™ Dura-Fit™ and Dura-Pak™ Shoe Covers
Catalog No. 19160240 Shop All Cardinal Health Products
Change view
Click to view available options
Description:
Dura-Fit™
Dura-Pak™
Sole Type:
Non-skid
Size:
2X-Large
Packaging Type:
100/Dispenser Box, 4 Boxes/Cs.
Bulk Packaged
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Sole Type Size Packaging Type
19-160-240 Dura-Pak™ Non-skid 2X-Large Bulk Packaged
19-160-235 Dura-Fit™ Non-skid 2X-Large 100/Dispenser Box, 4 Boxes/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-160-240 Supplier Cardinal Health™ Supplier No. 4875
This item is not returnable. View return policy
Made from strong, low linting SMS (spunbond meltblown spunbond) fabric. Available bulk packed in case of 200 pairs or packaged 50 pairs per dispenser box, four boxes per case.
  • Fluid resistant and 100% latex-free material
  • With or without skid-resistant tread
  • Blue
Order Info
Bulk packed 400/Cs. or dispenser boxes of 100/Box and 4 Boxes/Cs.
Specifications
Size 2X-Large
Material SMS
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Sole Type Non-skid
Description Dura-Pak™
Model 4875
Packaging Quantity 400/Cs.
Packaging Type Bulk Packaged
Product Type Shoe Covers