Learn More
MSC DuPont Hooded Coverall, Chemical Resistant, X-Large, 6 Pack
Catalog No. 5000782097
Encompass
DuPont™ Tychem® 6000 hooded coveralls are constructed from a proprietary barrier film laminated to a Tyvek® substrate, offering a broad chemical barrier against warfare agents and industrial chemicals. These gray coveralls provide at least 30 minutes of protection against over 180 challenge chemicals, ideal for military, law enforcement, and emergency response.
  • Provides 30+ minutes protection against 180+ challenge chemicals
  • Hooded coverall design with attached socks
  • Taped seams ensure strong chemical resistance against heavy liquid splashes
  • Three-piece respirator fit hood with elastic for complete neck coverage
  • Front zipper with semi-auto locking slider and double storm flap
  • Elastic wrists for a tighter fit
  • Integrated socks made of garment material
  • Meets ASTM F1670/F1671 for blood and viral penetration protection
Catalog No. 50-007-82097 Supplier MSC Supplier No. 02144988
DuPont™ Tychem® 6000 hooded coveralls are constructed from a proprietary barrier film laminated to a Tyvek® substrate, offering a broad chemical barrier against warfare agents and industrial chemicals. These gray coveralls provide at least 30 minutes of protection against over 180 challenge chemicals, ideal for military, law enforcement, and emergency response.
  • Provides 30+ minutes protection against 180+ challenge chemicals
  • Hooded coverall design with attached socks
  • Taped seams ensure strong chemical resistance against heavy liquid splashes
  • Three-piece respirator fit hood with elastic for complete neck coverage
  • Front zipper with semi-auto locking slider and double storm flap
  • Elastic wrists for a tighter fit
  • Integrated socks made of garment material
  • Meets ASTM F1670/F1671 for blood and viral penetration protection