Learn More
MSC DuPont Hooded Coverall, Microporous Film, Respirator Fit Hood, 25/Case
Catalog No. 5000676260
Encompass
The DuPont™ ProShield® 50 is a hooded coverall made from a lightweight microporous film garment. It is designed to offer protection against non-hazardous aerosols, light liquid splash, and dry particles. The white coverall features a respirator fit hood, elastic wrists and ankles, a storm flap, and a pin lock slider zipper pull. It is suitable for use in janitorial, sanitation, and light industrial applications.
  • Constructed from composite of microporous film and nonwoven fabric
  • Design includes serged seams, a respirator fit hood with elastic around the face, a front zipper closure with a storm flap, elastic wrists, and elastic ankles
  • White in color
  • Zipper features a pin locking slider that locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
Catalog No. 50-006-76260 Supplier MSC Supplier No. 38564563
The DuPont™ ProShield® 50 is a hooded coverall made from a lightweight microporous film garment. It is designed to offer protection against non-hazardous aerosols, light liquid splash, and dry particles. The white coverall features a respirator fit hood, elastic wrists and ankles, a storm flap, and a pin lock slider zipper pull. It is suitable for use in janitorial, sanitation, and light industrial applications.
  • Constructed from composite of microporous film and nonwoven fabric
  • Design includes serged seams, a respirator fit hood with elastic around the face, a front zipper closure with a storm flap, elastic wrists, and elastic ankles
  • White in color
  • Zipper features a pin locking slider that locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex