Learn More
MSC Dupont Hooded Coverall, Tychem 2000, 5X, Yellow, Serged Seams, 12/Case
Catalog No. 5000782295
Encompass
This hooded coverall provides lightweight and durable protection against a variety of industrial chemical threats. It is designed for light liquid splash protection in diverse industrial environments such as chemical processing, pharmaceutical manufacturing, and petroleum refining.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Serged seams
  • Pin lock slider zipper pull
  • Provides at least 30 minutes of protection against more than 40 chemical challenges
  • Manufactured without natural rubber latex components
  • Antistatic agent minimizes static build up
Catalog No. 50-007-82295 Supplier MSC Supplier No. 09694787
This hooded coverall provides lightweight and durable protection against a variety of industrial chemical threats. It is designed for light liquid splash protection in diverse industrial environments such as chemical processing, pharmaceutical manufacturing, and petroleum refining.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Serged seams
  • Pin lock slider zipper pull
  • Provides at least 30 minutes of protection against more than 40 chemical challenges
  • Manufactured without natural rubber latex components
  • Antistatic agent minimizes static build up