Learn More
MSC DuPont Hooded Coverall, Tychem 2000 Chemical Splash, 12/Pk.
Catalog No. 5000783051
Encompass
This Tychem 2000 hooded coverall provides lightweight, durable protection against various industrial chemical threats. Designed for light liquid splash protection, it is suitable for applications in chemical processing, petroleum refining, pharmaceutical manufacturing, and food processing.
  • Constructed from Tychem 2000 fabric (Tyvek coated with 1.25 mils of polyethylene)
  • Features attached hood with elastic face opening, elastic wrists, and elastic ankles
  • Equipped with serged seams and a storm flap with adhesive closure
  • Offers at least 30 minutes of protection against over 40 chemical challenges
  • Manufactured without natural rubber latex
  • Substrate treated with a topical antistatic agent
  • Meets or exceeds ANSI/ISEA 101 sizing standards
  • Usable in temperatures from -13°F (-25°C) to 165°F (74°C)
Catalog No. 50-007-83051 Supplier MSC Supplier No. 78726411
This Tychem 2000 hooded coverall provides lightweight, durable protection against various industrial chemical threats. Designed for light liquid splash protection, it is suitable for applications in chemical processing, petroleum refining, pharmaceutical manufacturing, and food processing.
  • Constructed from Tychem 2000 fabric (Tyvek coated with 1.25 mils of polyethylene)
  • Features attached hood with elastic face opening, elastic wrists, and elastic ankles
  • Equipped with serged seams and a storm flap with adhesive closure
  • Offers at least 30 minutes of protection against over 40 chemical challenges
  • Manufactured without natural rubber latex
  • Substrate treated with a topical antistatic agent
  • Meets or exceeds ANSI/ISEA 101 sizing standards
  • Usable in temperatures from -13°F (-25°C) to 165°F (74°C)