Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. 190881121B Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-1121B Medium
19-088-1121C Large
19-088-1121D X-Large
19-088-1121E 2X-Large
19-088-1121F 3X-Large
19-088-1121G 4X-Large
19-088-1121A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-088-1121B Supplier DuPont™ Supplier No. IC182BWHMD00250C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tyvek IsoClean delivers an ideal balance of protection, durability, and comfort. Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.

  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing.
  • Raglan sleeve design for greater range of motion and comfort.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Size (English) Chest 36 to 38 in.
Size (English) Inseam 32.5 in.
Size (Metric) Chest 91.44 to 96.52 cm
Size (Metric) Inseam 82.55 cm
Thickness 5.4 mil
For Use With (Application) Premium protection for controlled environments and inherent barrier to dry particles, microorganisms, and non-hazardous liquids
Length (English) Sleeve 37 in.
Length (Metric) Sleeve 93.98 cm
Height (English) 5.5 to 5.10 ft.
Height (Metric) 167.64 to 155.44 cm
Packaging Clean-processed
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.