Learn More
DuPont™ Tyvek™ IC668B IsoClean™ Full Face Opening Hoods
GREENER_CHOICE
Catalog No. 17988112 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Hood Type:
Clean-processed
Sterility:
Non-sterile
Catalog No. Hood Type Sterility
17-988-112 Clean-processed Non-sterile
1 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-988-112 Supplier DuPont™ Supplier No. IC668BWH0001000C
Add to Cart
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
IsoClean open face hoods provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Available in non-sterile, clean-processed, and sterile versions.
  • Bound seams are covered with garment fabric for reinforcement
  • Bound hood opening for lower particle shedding
  • Full face opening
  • Ties with loops for adjustable fit
  • One size fits most
  • Availiable in non-sterile, clean-processed, and sterile versions
  • Available in bulk packaging with double transparent poly liners
  • Clean-processed garments meet ISO Class Cleanroom 4 standards
  • Sterilzed hoods = gamma sterilzed to SAL of 10-6
TRUSTED_SUSTAINABILITY
Specifications
Product Type Full Face Opening
Size Universal
Material Tyvek
Color White
Closure Type Ties
Disposable Single-use
Model IC668B
Seam Type Bound
Sterility Non-sterile
Quantity 100/Cs.
Length (English) 17.25 in.
Length (Metric) 43.81 cm
Includes Ties with loops for fit
Hood Type Clean-processed
Cleanroom Class Class 1-10 (ISO 3-4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.