Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Frock
BIOPHARMAGREENER_CHOICE

Catalog No. 19161156 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-156 Small
19-088-1127B Medium
19-088-1127C Large
19-088-1127D X-Large
19-088-1127E 2X-Large
19-088-1127F 3X-Large
17-990-512 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19-161-156 Supplier DuPont™ Supplier No. IC270BWHSM0030CS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Clean-processed and sterile frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • Garments are gamma sterilized to SAL of 10-6.
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Full traceability on all sterilized apparel with Certificates of Sterility.
TRUSTED_SUSTAINABILITY

Specifications

Size Small
Material Tyvek
Color White
Disposable Single-use
Sterility Sterile
Closure Type Snaps
Cuff Style Elastic
No. of Pockets None
Style IsoClean™ 270 Series
Size (English) Chest 35 to 38 in.
Size (Metric) Chest 88 to 96 cm
Sleeve Style Long Sleeve
Packaging Type Clean-processed, Individually Packaged
Length (Metric) 99 cm
Length (English) Sleeve 34.75 in.
Length (Metric) Sleeve 88.3 cm
Cleanroom Class Class 100 (ISO 5)
Length (English) 39 in.
Product Type Controlled Environments Frock
Seam Type Bound
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.