Learn More
MSC DuPont Jacket, Tychem 5000, Tan, Hook-and-Loop Closure, Mandarin Collar, 6/Pk
Catalog No. 5000794238
Encompass
This chemical-resistant jacket provides durability for hazardous environments. It features a mandarin collar, jam fit cuff, double storm flap with hook and loop closure, and taped seams. Suitable for chemical handling, emergency response, and industrial cleaning.
  • Multi-layer film barrier laminated to durable polypropylene substrate
  • Taped seams offer strong chemical resistance against heavy liquid splashes
  • Mandarin collar designed for tight fit around hooded PAPR
  • Storm flap covers zipper, secured with hook and loop material
  • Jam fit cuff with glove inserts for user-supplied gloves
  • Manufactured without natural rubber latex (as of Jan 2023)
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
  • Protects against hazardous powders, particulates, and chemical spray
Catalog No. 50-007-94238 Supplier MSC Supplier No. 53841227
This chemical-resistant jacket provides durability for hazardous environments. It features a mandarin collar, jam fit cuff, double storm flap with hook and loop closure, and taped seams. Suitable for chemical handling, emergency response, and industrial cleaning.
  • Multi-layer film barrier laminated to durable polypropylene substrate
  • Taped seams offer strong chemical resistance against heavy liquid splashes
  • Mandarin collar designed for tight fit around hooded PAPR
  • Storm flap covers zipper, secured with hook and loop material
  • Jam fit cuff with glove inserts for user-supplied gloves
  • Manufactured without natural rubber latex (as of Jan 2023)
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
  • Protects against hazardous powders, particulates, and chemical spray