Learn More
MSC DuPont Jacket, Tychem 5000, Taped Seams, Tan, 6/Case
Catalog No. 5000802586
Encompass
This jacket offers heavy-duty protection against a wide range of chemical hazards. It features a mandarin collar for a tight fit around a hooded PAPR, elastic wrists, a double storm flap with hook and loop closure, and taped seams. It is ideal for chemical handling, hazmat operations, and domestic preparedness.
  • Strong, durable fabric provides barrier to a broad range of chemicals
  • Taped seams offer strong chemical resistance against heavy liquid splashes
  • Mandarin collar designed for tight fit around hooded PAPR
  • Double storm flap with hook and loop closure covers zipper
  • Elastic wrists for a secure fit
  • Manufactured without natural rubber latex components
  • Meets U.S. industry requirements for blood and viral penetration (ASTM F1670/F1671)
Catalog No. 50-008-02586 Supplier MSC Supplier No. 53841235
This jacket offers heavy-duty protection against a wide range of chemical hazards. It features a mandarin collar for a tight fit around a hooded PAPR, elastic wrists, a double storm flap with hook and loop closure, and taped seams. It is ideal for chemical handling, hazmat operations, and domestic preparedness.
  • Strong, durable fabric provides barrier to a broad range of chemicals
  • Taped seams offer strong chemical resistance against heavy liquid splashes
  • Mandarin collar designed for tight fit around hooded PAPR
  • Double storm flap with hook and loop closure covers zipper
  • Elastic wrists for a secure fit
  • Manufactured without natural rubber latex components
  • Meets U.S. industry requirements for blood and viral penetration (ASTM F1670/F1671)