Learn More
Netmercury DUPONT KRYTOX LVP GREASE 2OZ
Catalog No. NC9534760
Encompass
Krytox LVP 2oz high-vacuum grease performs over extremes of temperature and pressure that hydrocarbon, silicone, and chlorofluorocarbon greases can't match. From -15 to 300 deg.C (0-570 deg.F), at vapor pressures as low as 10-13 mmHg
Catalog No. NC9534760 Supplier Netmercury Supplier No. NM00020181
Krytox LVP 2oz high-vacuum grease performs over extremes of temperature and pressure that hydrocarbon, silicone, and chlorofluorocarbon greases can't match. From -15 to 300 deg.C (0-570 deg.F), at vapor pressures as low as 10-13 mmHg