Learn More
MSC Dupont Lab Coat, Tyvek, 3X-Large, 1.2 oz, 30 Pack
Catalog No. 5000795665
Encompass
The Dupont Tyvek 400 Lab Coat provides protective coverage for laboratory and industrial environments. Made from Tyvek material, this lab coat features a lightweight 1.2 oz construction and is designed for comfortable wear.
  • Size 3X-Large
  • Open cuff style
  • Snaps closure
  • Lightweight 1.20 oz material
  • Two pockets for convenience
  • Suitable for non-cleanroom applications
  • Made from durable Tyvek material
Catalog No. 50-007-95665 Supplier MSC Supplier No. 41550955
The Dupont Tyvek 400 Lab Coat provides protective coverage for laboratory and industrial environments. Made from Tyvek material, this lab coat features a lightweight 1.2 oz construction and is designed for comfortable wear.
  • Size 3X-Large
  • Open cuff style
  • Snaps closure
  • Lightweight 1.20 oz material
  • Two pockets for convenience
  • Suitable for non-cleanroom applications
  • Made from durable Tyvek material