Learn More
MSC Dupont Lab Coat, Tyvek 400, Chemical Resistant, Medium, 30/Pk.
Catalog No. 5000686812
Encompass
This lab coat is constructed from 1.20 oz Tyvek material, designed for chemical resistance. It features a medium size and snap closure, suitable for various laboratory applications.
  • Open cuff style
  • Material thickness of 5.90 mm
  • Non-cleanroom garment
  • Two pockets
  • Laydown collar
  • Sleeve length of 32 in
  • Meets standards ASTM D774, OSHA 16 CFR 1610, ASTM D257, ASTM D5034, AATCC 127, ASTM D1683, ASTM D3776, ASTM D1777
Catalog No. 50-006-86812 Supplier MSC Supplier No. 41550914
This lab coat is constructed from 1.20 oz Tyvek material, designed for chemical resistance. It features a medium size and snap closure, suitable for various laboratory applications.
  • Open cuff style
  • Material thickness of 5.90 mm
  • Non-cleanroom garment
  • Two pockets
  • Laydown collar
  • Sleeve length of 32 in
  • Meets standards ASTM D774, OSHA 16 CFR 1610, ASTM D257, ASTM D5034, AATCC 127, ASTM D1683, ASTM D3776, ASTM D1777