Learn More
MSC Dupont Lab Coat, Tyvek 400, Medium, 30/Pk
Catalog No. 5000768610
Encompass
This lab coat is constructed from Tyvek 400, a flash-spun, high-density polyethylene nonwoven material. It provides an inherent barrier against particles as small as 1.0 micron, offering protection, durability, and comfort for limited-use applications without films or laminates that can wear away.
  • Constructed from Tyvek 400 for protection against particles down to 1.0 micron
  • Features a body-length jacket design with front snap closures for easy donning and doffing
  • Includes a laydown collar and open cuffs
  • Offers resistance to hazardous dry particulates, non-hazardous light liquid splash, and aerosols
  • Suitable for applications such as asbestos abatement, general maintenance, and spray painting
  • Disposable design for limited-use applications
  • Size medium with a 32.25 in sleeve length
Catalog No. 50-007-68610 Supplier MSC Supplier No. 40600215
This lab coat is constructed from Tyvek 400, a flash-spun, high-density polyethylene nonwoven material. It provides an inherent barrier against particles as small as 1.0 micron, offering protection, durability, and comfort for limited-use applications without films or laminates that can wear away.
  • Constructed from Tyvek 400 for protection against particles down to 1.0 micron
  • Features a body-length jacket design with front snap closures for easy donning and doffing
  • Includes a laydown collar and open cuffs
  • Offers resistance to hazardous dry particulates, non-hazardous light liquid splash, and aerosols
  • Suitable for applications such as asbestos abatement, general maintenance, and spray painting
  • Disposable design for limited-use applications
  • Size medium with a 32.25 in sleeve length