Learn More
MSC Dupont Lab Coat, Tyvek 400, White, 4X-Large, 2 Pockets, 30/Case
Catalog No. 5000803088
Encompass
This lab coat is constructed from flash-spun high-density polyethylene, a unique nonwoven material that provides an inherent barrier against particles as small as 1.0 micron. It offers robust protection against fine particles and light liquid splash for various laboratory and maintenance applications.
  • Provides protection against fine particles and light liquid splash
  • Breathable and lightweight design
  • Inherent barrier against particles down to 1.0 micron
  • Protection built into the fabric for consistent barrier
  • Features front snap closures for easy donning
  • Equipped with a laydown collar and open wrists
  • Includes two front pockets
  • Constructed with serged seams
  • Manufactured without natural rubber latex
  • Treated with topical antistatic agent
Catalog No. 50-008-03088 Supplier MSC Supplier No. 68823210
This lab coat is constructed from flash-spun high-density polyethylene, a unique nonwoven material that provides an inherent barrier against particles as small as 1.0 micron. It offers robust protection against fine particles and light liquid splash for various laboratory and maintenance applications.
  • Provides protection against fine particles and light liquid splash
  • Breathable and lightweight design
  • Inherent barrier against particles down to 1.0 micron
  • Protection built into the fabric for consistent barrier
  • Features front snap closures for easy donning
  • Equipped with a laydown collar and open wrists
  • Includes two front pockets
  • Constructed with serged seams
  • Manufactured without natural rubber latex
  • Treated with topical antistatic agent