Learn More
MSC DuPont Lab Coat, Tyvek 400, White, Snap Closure, 1 ea
Catalog No. 5000686105
Encompass
The DuPont™ Tyvek® 400 Lab Coat offers robust protection against fine particles and light liquid splash. This breathable, lightweight garment extends below the hip, making it suitable for a variety of applications, including laboratory and pharmaceutical handling. Made from flash-spun high-density polyethylene, the Tyvek® 400 fabric provides an inherent barrier against particles down to 1.0 micron.
  • Front snap closures for easy donning and doffing
  • Enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding instruments or parts
  • Manufactured without natural rubber latex
Catalog No. 50-006-86105 Supplier MSC Supplier No. 53612073
The DuPont™ Tyvek® 400 Lab Coat offers robust protection against fine particles and light liquid splash. This breathable, lightweight garment extends below the hip, making it suitable for a variety of applications, including laboratory and pharmaceutical handling. Made from flash-spun high-density polyethylene, the Tyvek® 400 fabric provides an inherent barrier against particles down to 1.0 micron.
  • Front snap closures for easy donning and doffing
  • Enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding instruments or parts
  • Manufactured without natural rubber latex