Learn More
Cole-Parmer DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 50 Ft
Catalog No. NC3921155 Shop All Cole-Parmer Products
Encompass_Preferred
For transfer of high-purity fluids in pharmaceutical and biotechnological applications
  • Low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from organic plasticizers, phthalates or latex additives
  • Meets USP Class VI, FDA registered and ISO 9001:2015 requirements
  • This platinum-cured silicone DuPont Liveo Pharma-50 tubing is designed to transfer ultra-pure liquids in pharmaceutical and biotechnological processes where contamination is a concern. The tubing is stable over a wide temperature range, is highly resilient, impairs no taste or odor, and offers a non-wetting surface. Made from BioMedical Grade elastomer, the tubing exceeds United States Pharmacopeia (USP) Class VI Plastics Test Requirements. It also meets the European Pharmacopoeia monograph 3.1.9. “Silicone elastomer for closures and tubing.” Easily sterilizable by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 50 kGy.
    Catalog No. NC3921155 Supplier Cole-Parmer Supplier No. EW5012192
    For transfer of high-purity fluids in pharmaceutical and biotechnological applications
  • Low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from organic plasticizers, phthalates or latex additives
  • Meets USP Class VI, FDA registered and ISO 9001:2015 requirements
  • This platinum-cured silicone DuPont Liveo Pharma-50 tubing is designed to transfer ultra-pure liquids in pharmaceutical and biotechnological processes where contamination is a concern. The tubing is stable over a wide temperature range, is highly resilient, impairs no taste or odor, and offers a non-wetting surface. Made from BioMedical Grade elastomer, the tubing exceeds United States Pharmacopeia (USP) Class VI Plastics Test Requirements. It also meets the European Pharmacopoeia monograph 3.1.9. “Silicone elastomer for closures and tubing.” Easily sterilizable by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 50 kGy.