Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 60 Hooded Coverall

Catalog No. 190882159 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-2159 Medium
19-088-2158 Small
19-088-2160 Large
19-088-2161 X-Large
19-088-2162 2X-Large
19-088-2163 3X-Large
19-088-2164 4X-Large
19-130-270 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-088-2159 Supplier DuPont™ Supplier No. NG127SWHMD0025NP
This item is not returnable. View return policy

Coveralls with respirator fit hood are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Liquid
Certifications/Compliance ASTM D751, D1117, D5034, D257, AATCC 127
Description Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Latex Free Yes
Model NG127S
Size (English) Chest 33.75 to 37.25 in.
Size (English) Inseam 26 in.
Size (Metric) Chest 85.72 to 94.61 cm
Size (Metric) Inseam 66.04 cm
Thickness 10 mil
For Use With (Application) Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation
Length (English) Sleeve 31.75 in.
Length (Metric) Sleeve 80.64 cm
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.