Learn More
DuPont™ ProShield™ 2 Coveralls
Catalog No. 19001834 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Elasticized wrists and ankles; Large
Catalog No.
19-001-834 Elasticized wrists and ankles; Large
1 options
This item is not returnable. View return policy
Catalog No. 19-001-834 Supplier DuPont™ Supplier No. P2132SWHLG0025ZF
This item is not returnable. View return policy
High barrier, high comfort
  • ProShield 2 provides an anti-static particulate and liquid barrier.
  • Made of nonwoven fabric and microporous film
  • Used in chemical handling and agricultural chemical applications.
  • Strong and tear resistant.
  • Treated to provide antistatic properties.
  • Minimum 26 in. (66 cm) zipper
  • Hooded styles zip to above the chin, eliminating exposed areas when wearer is using breathing apparatus.
  • Weight: 1.7 oz. per sq. yd.
  • Elastic back waist.
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Proprietary Bilaminate Composite Fabric
Color White
Disposable Single-use
Sterility Non-sterile