Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 3 Boot Covers (18 in. Tall)

Catalog No. 19715370 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Length: 16 in.; Size: X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-715-370 Length: 16 in.; Size: X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-715-370 Supplier DuPont™ Supplier No. P3454SGY00010000
This item is not returnable. View return policy

Provides coverage up to calf

  • 18 in. high boot cover with elastic top
  • ProShield 3 sole material provides enhanced skid-resistance
  • One-size-fits-most
  • Provides non-hazardous liquid splash protection

TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material Polypropylene
Color Gray
Disposable LDPE
Closure Type Elastic Top
Sole Type Non-skid
Seam Type Serged
Length (English) 16 in.
Length (Metric) 40.64 cm
Height (English) 18 in.
Height (Metric) 45.72 cm
Product Type Boot Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.