Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 10 Coverall (all)

Catalog No. 19162648 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-162-648 White 7X-Large
19-162-500 White Medium
19-162-501 White Large
19-162-503 White 2X-Large
19-162-502 White X-Large
19-162-504 White 3X-Large
19-162-505 White 4X-Large
19-162-506 White 5X-Large
19-162-510 Blue 2X-Large
19-162-511 Blue 3X-Large
19-162-507 Blue Medium
19-162-509 Blue X-Large
19-162-508 Blue Large
19-162-513 Blue 5X-Large
19-162-512 Blue 4X-Large
19-162-641 Blue 6X-Large
19-162-642 White 6X-Large
19-162-647 Blue 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 19-162-648 Supplier DuPont™ Supplier No. PB120SWH7X002500
  • $218.99
    $113.65 / Case of 25

    Save $105.34

    48% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 7X-Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Non-toxic Particulates
Collar Type Laydown
Model PB120S
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.