Learn More
DuPont™ ProShield™ 10 Lab Coat
Catalog No. 19162632 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-162-632 White 4X-Large
19-162-619 Blue Small
19-162-620 Blue Medium
19-162-621 Blue Large
19-162-622 Blue X-Large
19-162-623 Blue 2X-Large
19-162-624 Blue 3X-Large
19-162-625 Blue 4X-Large
19-162-626 White Small
19-162-627 White Medium
19-162-628 White Large
19-162-629 White X-Large
19-162-630 White 2X-Large
19-162-631 White 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-162-632 Supplier DuPont™ Supplier No. PB219SWH4X003000
  • $198.00
    $147.65 / Case of 30
    Save $50.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
SMS disposable lab coats are designed for a range of industries for nonhazardous dry particle and light liquid splash applications. Available in blue or white in a variety of sizes.
  • Knit collar
  • Set sleeve
  • Knit cuff
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
Style Standard Length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Model PB219
Seam Type Serged
Certifications/Compliance ASTM D1777