Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ NexGen™ Shoe Covers

Catalog No. 19715311 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-715-311 Medium
19-715-312 Large
19-715-313 X-Large
19-715-310 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-715-311 Supplier DuPont™ Supplier No. NG451SWHMD020000
This item is not returnable. View return policy

DuPont ProShield 60 Shoe Cover with PVC Skid-Resistant Sole. 5 in. High. Serged Seams.

ProShield 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications. ProShield 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include: automotive refinishing, waste cleanup and remediation.5 in. shoe cover with elastic top PVC soles provides enhanced skid-resistance and durability. One size fits most.
TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material Polypropylene
Color White
Closure Type Elastic Ankle
Sole Type Non-skid
Model NG451SWHMD020000
Seam Type Serged
Thickness 10 mil
For Use With (Application) Ideal for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications and automotive refinishing, waste cleanup, remediation
Length (English) 11.75 in.
Length (Metric) 29.84 cm
Height (English) 5 in.
Height (Metric) 12.7 cm
Resistance Dry Particulates, Fluid
Product Type Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.